accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COOHNOOCH3

CAS RN 31954-27-5

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: PHUZOEOLWIHIKH-UHFFFAOYSA-N
SMILES: COC(=O)CNC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)