accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31963-35-6

ONH2HCl

CAS RN 31963-35-6

IUPAC Name: 1-(2-phenoxyphenyl)methanamine hydrochloride
Synonyms: 1-(2-phenoxyphenyl)methanamine hydrochloride
Molecular Weight (g/mol): 235.71
Molecular Formula: C13H14ClNO
InChi Key: USRYZTSPSJXQFU-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=CC=C1OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results