accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 320-50-3

FFFClCl

CAS RN 320-50-3

IUPAC Name: 1,4-dichloro-2-(trifluoromethyl)benzene
Synonyms: 1,4-dichloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 215
Molecular Formula: C7H3Cl2F3
InChi Key: DYBYUWVMLBBEMA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results