accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClFFF

CAS RN 320-51-4

IUPAC Name: 4-chloro-3-(trifluoromethyl)aniline
Synonyms: ACBT
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: ASPDJZINBYYZRU-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)