accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHOOF

CAS RN 32004-66-3

IUPAC Name: 2-(5-fluoro-2-methyl-1H-inden-3-yl)acetic acid
Synonyms: (6-fluoro-2-methyl-3H-inden-1-yl)acetic acid
Molecular Weight (g/mol): 206.22
Molecular Formula: C12H11FO2
InChi Key: QDDPPRDVFIJASZ-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C2=CC(F)=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results