accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOPd(E)(E)(E)(E)

CAS RN 32005-36-0

IUPAC Name: bis((1E,4E)-1,5-diphenylpenta-1,4-dien-3-one) palladium
Synonyms: bis(dibenzylideneacetone) palladium
Molecular Weight (g/mol): 575.02
Molecular Formula: C34H28O2Pd
InChi Key: UKSZBOKPHAQOMP-SVLSSHOZSA-N
SMILES: [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)