accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32025-65-3

IUPAC Name: 2-(3-methoxyphenyl)-2-oxoacetaldehyde
Synonyms: 2-(3-methoxyphenyl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: SIHYQEYAJMDKQH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(=O)C=O
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results