accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3209-13-0

IUPAC Name: 3-methoxy-5-methylphenol
Synonyms: 5-methoxy-M-cresol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: NOTCZLKDULMKBR-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results