accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 321309-26-6

IUPAC Name: 5-methyl-1,2-oxazole-4-sulfonyl chloride
Synonyms: 5-methyl-1,2-oxazole-4-sulfonyl chloride
Molecular Weight (g/mol): 181.59
Molecular Formula: C4H4ClNO3S
InChi Key: STYUNYVEEFPQMF-UHFFFAOYSA-N
SMILES: CC1=C(C=NO1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results