accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 321309-28-8

IUPAC Name: 6-fluoro-2,4-dihydro-1,3-benzodioxine-8-carboxylic acid
Synonyms: 6-fluoro-2,4-dihydro-1,3-benzodioxine-8-carboxylic acid
Molecular Weight (g/mol): 198.15
Molecular Formula: C9H7FO4
InChi Key: HWBALMSPYAUMMB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC2=C1OCOC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results