accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 321309-32-4

IUPAC Name: 1,2,3-benzothiadiazole-5-carbonyl chloride
Synonyms: 1,2,3-benzothiadiazole-5-carbonyl chloride
Molecular Weight (g/mol): 198.62
Molecular Formula: C7H3ClN2OS
InChi Key: VOSTUOLSYXVSND-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C2SN=NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results