accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32137-76-1

H3COOSN

CAS RN 32137-76-1

IUPAC Name: ethyl 1,3-benzothiazole-2-carboxylate
Synonyms: ethyl 1,3-benzothiazole-2-carboxylate
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: VLQLCEXNNGQELL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results