accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOClNOO

CAS RN 321531-57-1

IUPAC Name: 4-chloro-3-nitro-N-propylbenzamide
Synonyms: 4-chloro-3-nitro-N-propylbenzamide
Molecular Weight (g/mol): 242.66
Molecular Formula: C10H11ClN2O3
InChi Key: ZGGRHAZHLVGWPF-UHFFFAOYSA-N
SMILES: CCCNC(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results