accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2N

CAS RN 3218-02-8

IUPAC Name: 1-cyclohexylmethanamine
Synonyms: (aminomethyl)cyclohexane
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: AVKNGPAMCBSNSO-UHFFFAOYSA-N
SMILES: NCC1CCCCC1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results