accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

TeTe

CAS RN 32294-60-3

IUPAC Name: (phenylditellanyl)benzene
Synonyms: diphenyl ditelluride
Molecular Weight (g/mol): 409.41
Molecular Formula: C12H10Te2
InChi Key: VRLFOXMNTSYGMX-UHFFFAOYSA-N
SMILES: [Te]([Te]C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results