accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32333-53-2

IUPAC Name: 4-chloro-3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 279.05
Molecular Formula: C7H3Cl2F3O2S
InChi Key: SSULGNXFUGLULI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=CC(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)