accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NbOOOOOH3CCH3H3CCH3CH3

CAS RN 3236-82-6

IUPAC Name: niobium(5+) pentaethanolate
Synonyms: niobium(V) ethoxide
Molecular Weight (g/mol): 318.21
Molecular Formula: C10H25NbO5
InChi Key: ZTILUDNICMILKJ-UHFFFAOYSA-N
SMILES: [Nb+5].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results