accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOOHO

CAS RN 3238-40-2

IUPAC Name: furan-2,5-dicarboxylic acid
Synonyms: 2,5-furandicarboxylic acid furane-α,α'-dicarboxylic acid dehydromucate furane-a,a'-dicarboxylic acid furan-2,5-dicarbonsaeure dehydromucic acid furane-a,a'-dicarboxylate dehydroschleimsaeure
Molecular Weight (g/mol): 156.09
Molecular Formula: C6H4O5
InChi Key: CHTHALBTIRVDBM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results