accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 324-74-3

IUPAC Name: 4-fluoro-1,1'-biphenyl
Synonyms: 4-fluorobiphenyl
Molecular Weight (g/mol): 172.2
Molecular Formula: C12H9F
InChi Key: RUYZJEIKQYLEGZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results