accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BK

CAS RN 3244-41-5

IUPAC Name: potassium tetraphenylboranuide
Synonyms: potassium tetraphenylborate
Molecular Weight (g/mol): 358.33
Molecular Formula: C24H20BK
InChi Key: MAMCUZQNCMYPCY-UHFFFAOYSA-N
SMILES: [K+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results