CAS RN 32446-66-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4,4'-Dicyanobenzophenone, TRC
CAS: 32446-66-5 Molecular Formula: C15 H8 N2 O Molecular Weight (g/mol): 232.24 Synonym: Benzonitrile, 4,4'-carbonylbis-,Benzonitrile, 4,4'-carbonyldi- (8CI),4,4'-Carbonylbis[benzonitrile],4,4'-Dicyanobenzophenone,4-[(4-Cyanophenyl)carbonyl]benzonitrile,Bis(4-cyanophenyl) ketone,Bis(p-cyanophenyl) ketone,p,p'-Dicyanobenzophenone IUPAC Name: 4-(4-cyanobenzoyl)benzonitrile SMILES: O=C(c1ccc(cc1)C#N)c2ccc(cc2)C#N
2
4,4'-Carbonyldibenzonitrile (4,4'-Dicyanobenzophenone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.