accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32499-64-2

IUPAC Name: ethyl (1R,5S)-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Synonyms: ethyl (1R,5S)-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Molecular Weight (g/mol): 197.23
Molecular Formula: C10H15NO3
InChi Key: ANEJUHJDPGTVIO-OCAPTIKFSA-N
SMILES: CCOC(=O)N1[C@H]2CC[C@@H]1CC(=O)C2
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)