accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 325129-69-9

BOOCH3CH3CH3CH3H3CCH3BOOH3CH3CH3CH3C

CAS RN 325129-69-9

IUPAC Name: 2-[9,9-dimethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[9,9-dimethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 446.2
Molecular Formula: C27H36B2O4
InChi Key: RVFLMSKITNJVRB-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(C=C(C=C1)B1OC(C)(C)C(C)(C)O1)C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results