accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNCH3CH3

CAS RN 325142-87-8

IUPAC Name: N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 247.15
Molecular Formula: C14H22BNO2
InChi Key: BIJRPONWUBCGES-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results