accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBrN

CAS RN 3252-43-5

IUPAC Name: 2,2-dibromoacetonitrile
Synonyms: dibromoacetonitrile
Molecular Weight (g/mol): 198.85
Molecular Formula: C2HBr2N
InChi Key: NDSBDLSWTGLNQA-UHFFFAOYSA-N
SMILES: BrC(Br)C#N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)