accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32597-34-5

OHNHN(E)

CAS RN 32597-34-5

IUPAC Name: N-{[(2E)-2H-pyrrol-2-ylidene]methyl}hydroxylamine
Synonyms: N-[(2E)-pyrrol-2-ylidenemethyl]hydroxylamine
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: WJMIDGRLWRUSSN-SNAWJCMRSA-N
SMILES: ON\C=C1/C=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results