CAS RN 32634-68-7
IUPAC Name:
(2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Synonyms:
(2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular Weight (g/mol):
386.36
Molecular Formula:
C20H18O8
InChi Key:
CMIBUZBMZCBCAT-HOTGVXAUSA-N
SMILES:
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O