accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O

CAS RN 3264-21-9

IUPAC Name: 1-(pyren-1-yl)ethan-1-one
Synonyms: 1-acetylpyrene
Molecular Weight (g/mol): 244.29
Molecular Formula: C18H12O
InChi Key: KCIJNJVCFPSUBQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)