accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFF

CAS RN 327-54-8

IUPAC Name: 1,2,4,5-tetrafluorobenzene
Synonyms: 1,2,4,5-tetrafluorobenzene
Molecular Weight (g/mol): 150.08
Molecular Formula: C6H2F4
InChi Key: SDXUIOOHCIQXRP-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C=C1F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results