accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFClNCO

CAS RN 327-78-6

IUPAC Name: 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
Synonyms: 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 221.56
Molecular Formula: C8H3ClF3NO
InChi Key: NBJZEUQTGLSUOB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC=C1Cl)N=C=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)