accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 327056-62-2

NFN

CAS RN 327056-62-2

IUPAC Name: 5-fluoropyridine-2-carbonitrile
Synonyms: 5-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: BHXHRMVSUUPOLX-UHFFFAOYSA-N
SMILES: FC1=CN=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results