accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32780-06-6

IUPAC Name: 5-(hydroxymethyl)oxolan-2-one
Synonyms: 5-(hydroxymethyl)oxolan-2-one
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: NSISJFFVIMQBRN-UHFFFAOYNA-N
SMILES: OCC1CCC(=O)O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)