accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3HNOHOHH2NO

CAS RN 32780-64-6

IUPAC Name: 2-hydroxy-5-{1-hydroxy-2-[(4-phenylbutan-2-yl)amino]ethyl}benzamide hydrochloride
Synonyms: labetalol hydrochloride
Molecular Weight (g/mol): 364.87
Molecular Formula: C19H25ClN2O3
InChi Key: WQVZLXWQESQGIF-UHFFFAOYNA-N
SMILES: Cl.CC(CCC1=CC=CC=C1)NCC(O)C1=CC=C(O)C(=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results