accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 328-69-8

FFFHOOOH

CAS RN 328-69-8

IUPAC Name: 3-hydroxy-5-(trifluoromethyl)benzoic acid
Synonyms: 3-hydroxy-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: BJUOAPFXYPEEMK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results