accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFFFNOO

CAS RN 328-80-3

IUPAC Name: 3-nitro-5-(trifluoromethyl)benzoate
Synonyms: 3-nitro-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 234.11
Molecular Formula: C8H3F3NO4
InChi Key: ODCLHXGXGFBBTA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=CC(=C1)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results