accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3282-11-9

ONONOO

CAS RN 3282-11-9

IUPAC Name: 4-nitro-4'-(4-nitrophenyl)-1,1'-biphenyl
Synonyms: 4-nitro-4'-(4-nitrophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 320.3
Molecular Formula: C18H12N2O4
InChi Key: MHOAYDHUNGLDTB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results