accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32874-26-3

H3COOCOCOCOCr

CAS RN 32874-26-3

IUPAC Name: ethyl benzoate tris(methanidylidyneoxidanium) chromium
Synonyms: tris(carbon monoxide) ethyl benzoate chromium
Molecular Weight (g/mol): 286.2
Molecular Formula: C12H10CrO5
InChi Key: SWSAJUVURPSGIG-UHFFFAOYSA-N
SMILES: [Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].CCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results