accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32890-88-3

IUPAC Name: 2,6-difluoro-3-methylbenzoic acid
Synonyms: 2,6-difluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: QZIVNVIPFGKDQE-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results