accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOOHClCl

CAS RN 32890-91-8

IUPAC Name: 2,3-dichloro-6-fluorobenzoic acid
Synonyms: 2,3-dichloro-6-fluorobenzoic acid
Molecular Weight (g/mol): 209
Molecular Formula: C7H3Cl2FO2
InChi Key: OOQKLNYJVMPYIF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results