accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 329-01-1

IUPAC Name: 1-isocyanato-3-(trifluoromethyl)benzene
Synonyms: 1-isocyanato-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: SXJYSIBLFGQAND-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)N=C=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)