accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 329-15-7

IUPAC Name: 4-(trifluoromethyl)benzoyl chloride
Synonyms: 4-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: OXZYBOLWRXENKT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results