accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32916-51-1

IUPAC Name: magnesium(2+) cyclopentanide chloride
Synonyms: magnesium(2+) cyclopentanide chloride
Molecular Weight (g/mol): 128.88
Molecular Formula: C5H9ClMg
InChi Key: KQIVBIRNYZIHNE-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C1CC[CH-]C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results