accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COBOH3CH3CN

CAS RN 329214-79-1

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 205.06
Molecular Formula: C11H16BNO2
InChi Key: XEMDFESAXKSEGI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CN=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results