accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CCH3CCH3CY

CAS RN 329735-72-0

IUPAC Name: yttrium(3+) tris(2-methylcyclopenta-1,3-dien-1-ide)
Synonyms: yttrium(3+) tris(2-methylcyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol): 326.27
Molecular Formula: C18H21Y
InChi Key: OXKRLQPVIZJLDT-UHFFFAOYSA-N
SMILES: [Y+3].CC1=[C-]CC=C1.CC1=[C-]CC=C1.CC1=[C-]CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results