accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32981-85-4

IUPAC Name: methyl (2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoate
Synonyms: methyl (2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoate
Molecular Weight (g/mol): 299.33
Molecular Formula: C17H17NO4
InChi Key: UYJLJICUXJPKTB-LSDHHAIUSA-N
SMILES: COC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results