accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 330-13-2

IUPAC Name: (4-nitrophenoxy)phosphonic acid
Synonyms: 4-nitrophenyl phosphate nitrophenylphosphate para-nitrophenylphosphate
Molecular Weight (g/mol): 219.09
Molecular Formula: C6H6NO6P
InChi Key: XZKIHKMTEMTJQX-UHFFFAOYSA-N
SMILES: OP(O)(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results