accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOSFFF

CAS RN 330-17-6

IUPAC Name: 4-[(trifluoromethyl)sulfanyl]benzoate
Synonyms: 4-[(trifluoromethyl)sulfanyl]benzoate
Molecular Weight (g/mol): 221.17
Molecular Formula: C8H4F3O2S
InChi Key: UMOGQQWVQUQTQA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(SC(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results