accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 330-55-2

IUPAC Name: 1-(3,4-dichlorophenyl)-3-methoxy-3-methylurea
Synonyms: linuron (iso)
Molecular Weight (g/mol): 249.09
Molecular Formula: C9H10Cl2N2O2
InChi Key: XKJMBINCVNINCA-UHFFFAOYSA-N
SMILES: CON(C)C(=O)NC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Solvent or Matrix 
  • (1)
  • (2)
Concentration 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)