accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33008-06-9

H3CNCH3SOOHNNH2

CAS RN 33008-06-9

IUPAC Name: 5-(dimethylamino)naphthalene-1-sulfonohydrazide
Synonyms: 5-(dimethylamino)naphthalene-1-sulfonohydrazide
Molecular Weight (g/mol): 265.33
Molecular Formula: C12H15N3O2S
InChi Key: KPQYDVAFRDWIBW-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NN
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results