accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3303-84-2

H3CCH3H3COONHOHO

CAS RN 3303-84-2

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: 3-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 188.2
Molecular Formula: C8H14NO4
InChi Key: WCFJUSRQHZPVKY-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)